Learn More
DLK1 Mouse anti-Human, Unconjugated, Clone: 2C3H8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67923-1-IG-20UL
This item is not returnable.
View return policy
Description
The DLK1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human DLK1. This antibody recognizes human, pig antigen. The DLK1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DLK1 | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 8788 | |
| IgG1 |
| Western Blot, ELISA | |
| 2C3H8 | |
| Mouse | |
| Primary | |
| Human, Pig |