missing translation for 'onlineSavingsMsg'
Learn More

DLK-IN-1, MedChemExpress

Product Code. p-200099268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487557 5 mg 5mg
16475606 100 mg 100mg
16431077 50 mg 50mg
16423256 25 mg 25mg
16404818 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487557 Supplier Medchem Express Supplier No. HY114331
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DLK-IN-1 is a selective, orally active inhibitor of dual leucine zipper kinase (DLK, MAP3K12), with a Ki of 3 nM. DLK-IN-1 retains excellent CNS penetration and is well tolerated following multiple days of dosing at concentrations that exceed those required for DLK inhibition in the brain. DLK-IN-1 has activity in a model of Alzheimer’s Disease.

DLK-IN-1 is a selective, orally active inhibitor of dual leucine zipper kinase (DLK, MAP3K12), with a Ki of 3 nM. DLK-IN-1 retains excellent CNS penetration and is well tolerated following multiple days of dosing at concentrations that exceed those required for DLK inhibition in the brain. DLK-IN-1 has activity in a model of Alzheimer’s Disease.

Specifications

Chemical Name or Material DLK-IN-1
CAS 1620574-24-4
Color White
Purity Grade Notes Research
Molecular Formula C20H24F3N5O2
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (147.60 mM; Need ultrasonic)
SMILES CC(N1C([C@@H]2[C@]3([H])[C@@]2([H])CN(C4COC4)C3)=CC(C5=CN=C(N)C(OC(F)(F)F)=C5)=N1)C
Molecular Weight (g/mol) 423.43
Formula Weight 423.43
Percent Purity 99.41%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.