Learn More
DLin-KC2-DMA, MedChemExpress
Description
DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.
Specifications
Specifications
| Chemical Name or Material | DLin-KC2-DMA |
| CAS | 1190197-97-7 |
| Color | Colorless |
| Purity Grade Notes | Research |
| Molecular Formula | C43H79NO2 |
| Quantity | 10 mg |
| Solubility Information | Ethanol : 100 mg/mL (155.74 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (155.74 mM; Need ultrasonic) |
| SMILES | CCCCC/C=C\C/C=C\CCCCCCCCC1(CCCCCCCC/C=C\C/C=C\CCCCC)OC(CCN(C)C)CO1 |
| Molecular Weight (g/mol) | 642.09 |
| Formula Weight | 642.09 |
| Show More |