Learn More
Invitrogen™ DLD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579156
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human A431 whole cell, human SiHa whole cell, human U251 whole cell, rat liver tissue, rat heart tissue, mouse liver tissue, mouse heart tissue. IHC: Human Mammary Cancer Tissue. ICC/IF: HCT116 cell, MCF-7 Cell.
This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
Specifications
| DLD | |
| Polyclonal | |
| Unconjugated | |
| Dld | |
| aei; after eight protein; branched chain alpha-keto acid dehydrogenase complex subunit E3; deltaD; deltaD transmembrane protein; delta-like protein D; diaphorase; dihydrolipoamide dehydrogenase; dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex); dihydrolipoamide: NAD+ oxidoreductase; dihydrolipoyl dehydrogenase mitochondrial-like protein; dihydrolipoyl dehydrogenase, mitochondrial; DLD; DLDD; DLDH; E3; E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex; GCSL; Glycine cleavage system L protein; glycine cleavage system protein L; LAD; lipoamide dehydrogenase; lipoamide dehydrogenase precursor; lipoamide reductase; lipoyl dehydrogenase; LOW QUALITY PROTEIN: dihydrolipoyl dehydrogenase, mitochondrial; mitochondrial Dihydrolipoyl dehydrogenase-like protein; OTTHUMP00000206744; OTTHUMP00000206746; OTTHUMP00000206748; OTTHUMP00000206749; PH; PHE 3; PHE3; QflA-16416; QtsA-11266; sc:d0402; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13382, 1738, 298942 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.01mg sodium azide, 0.01mg thimerosal | |
| O08749, P09622, Q6P6R2 | |
| Dld | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human DLD (492-509aa EAFREANLAASFGKSINF). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.