Learn More
Thermo Scientific Chemicals DL-Methionine sulfoxide, 98+%
Product Code. p-7023978
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449226 5 g 5g
11459226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11449226 Supplier Thermo Scientific Chemicals Supplier No. A18081.06
Request Bulk or Custom Format
CAS: 62697-73-8 | C5H11NO3S | 165.21 g/mol
DL-Methionine sulfoxide is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Methionine sulfoxide is used as pharmaceutical intermediates.
Solubility
Soluble in water and Hcl.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62697-73-8
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00002620
InChI Key QEFRNWWLZKMPFJ-UHFFFAOYNA-N
Synonym dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinyl
PubChem CID 847
ChEBI CHEBI:49033
SMILES CS(=O)CCC(N)C(O)=O
Specifications
Melting Point ∼230°C (decomposition)
Solubility Information Soluble in water and Hcl.
IUPAC Name 2-amino-4-methylsulfinylbutanoic acid
Quantity 5 g
Formula Weight 165.21
Percent Purity ≥98%
Chemical Name or Material DL-Methionine sulfoxide