missing translation for 'onlineSavingsMsg'
Learn More

dl-Mandelic Acid, Sigma-Aldrich™

Product Code. 15834201
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15834201 1 kg 1kg
15824201 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15834201 Supplier Sigma Aldrich Supplier No. 8069131000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-64-2

Specifications

Melting Point 119°C to 121°C
Density 1.31 g/cm3 at 20.2°C
Quantity 1 kg
Molecular Formula C6H5CH(OH)COOH
MDL Number MFCD00064250
Synonym DL-Mandelic acid, DL-α-Hydroxyphenylacetic acid, Phenylglycollic acid, DL-Mandelic acid
Molecular Weight (g/mol) 152.15
Grade Ungraded
Chemical Name or Material DL-Mandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.