missing translation for 'onlineSavingsMsg'
Learn More

dl-Mandelic Acid Nitrile, Sigma-Aldrich™

Product Code. 15894471
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894471 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15894471 Supplier Sigma Aldrich Supplier No. 8190160100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 532-28-5
Melting Point -10°C
Boiling Point 170°C/1013 hPa
Density 1.121 To 1.127 g/cm3 at 20°C
Quantity 100 mL
MDL Number MFCD00004487
Synonym DL-Mandelic acid nitrile, Benzaldehyde cyanohydrin, DL-α-Hydroxyphenylacetonitrile, DL-Mandelic acid nitrile
Molecular Weight (g/mol) 133.15
Chemical Name or Material DL-Mandelic Acid Nitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.