Learn More
DL-Malic acid, 98%
Description
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.
Solubility
Soluble in acetone, ether, water, methanol and ethanol.
Notes
Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
Specifications
Specifications
| Melting Point | 130°C to 133°C |
| Assay Percent Range | 0.98 |
| Beilstein | 1723539 |
| Solubility Information | Soluble in acetone,ether,water,methanol and ethanol. |
| Formula Weight | 134.09 |
| Density | 1.609 |
| Quantity | 500 g |
| Flash Point | 203°C (397°F) |
| Chemical Name or Material | DL-Malic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.