Learn More
dl-Limonene, Sigma-Aldrich™
Product Code. 15886520
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886520 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15886520 Supplier Sigma Aldrich Supplier No. 8145461000
 Restricted Item
Estimated Shipment: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
Mixture of D- and L-form ∼1:1 for synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 138-86-3
Melting Point -89°C
Density 0.84 g/cm3 at 20°C
Boiling Point 178°C/1013 hPa
MDL Number MFCD00062992
Quantity 1 L
Synonym DL-Limonene, DL-p-Mentha-1,8-diene, 4-Isopropyl-1-methylcyclohexene, DL-Limonene
Molecular Weight (g/mol) 136.23
Grade Research