Learn More
Thermo Scientific Chemicals DL-Leucine, 99+%
Product Code. p-153651 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
25g
Product Code. 10741641
Brand: Thermo Scientific Chemicals 172130250
Request Bulk or Custom Format
CAS: 328-39-2 | C6H13NO2 | 131.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 328-39-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063087
InChI Key ROHFNLRQFUQHCH-UHFFFAOYNA-N
Synonym dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d
PubChem CID 857
ChEBI CHEBI:25017
IUPAC Name 2-amino-4-methylpentanoic acid
SMILES CC(C)CC(N)C(O)=O
Specifications
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.3% max.
Packaging Plastic Bottle
Linear Formula (CH3)2CHCH2CH(NH2)CO2H
Beilstein 04, 447
Merck Index 14, 5451
Sulfated Ash 0.1% max.
Quantity 25 g
Formula Weight 131.17
Percent Purity 99.0 to 101.0% (on dry substance)
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Leucine, 99+%