Learn More
DL-Cysteine Hydrochloride Hydrate, TRC
Product Code. 30361226
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361226 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30361226 Supplier Toronto Research Chemicals Supplier No. TRCC99502550G
 Restricted Item
Estimated Shipment: 26-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DL-Cysteine Hydrochloride Hydrate
CAS 96998-61-7
Molecular Formula C3H7NO2S .HCl .xH2O
Quantity 50 g
Synonym Cysteine Hydrochloride Hydrate
InChI Formula InChI=1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2
SMILES SCC(N)C(O)=O.Cl.[H2O]
IUPAC Name cysteine hydrochloride hydrate
Molecular Weight (g/mol) 175.64