missing translation for 'onlineSavingsMsg'
Learn More

DL-beta-Proline hydrochloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7038411
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
953079-94-2
C5H10ClNO2
151.59
MFCD06801272
OYCLYMMIZJWYJG-UHFFFAOYSA-N
pyrrolidine-3-carboxylic acid hydrochloride, beta-proline hcl, 3-pyrrolidinecarboxylic acid hcl, 3-pyrrolidinecarboxylic acid hydrochloride, pubchem16223, beta-proline hydrochloride, ksc494m2f, pyrrolidine-3-carboxylicacidhydrochloride, 3-pyrrolidine carboxylic acid hydrochloride, rs-pyrrolidine-3-carboxylic acid hydrochloride
44890816
pyrrolidine-3-carboxylic acid;hydrochloride
C1CNCC1C(=O)O.Cl
This item is not returnable. View return policy
953079-94-2
C5H10ClNO2
151.59
MFCD06801272
OYCLYMMIZJWYJG-UHFFFAOYSA-N
pyrrolidine-3-carboxylic acid hydrochloride, beta-proline hcl, 3-pyrrolidinecarboxylic acid hcl, 3-pyrrolidinecarboxylic acid hydrochloride, pubchem16223, beta-proline hydrochloride, ksc494m2f, pyrrolidine-3-carboxylicacidhydrochloride, 3-pyrrolidine carboxylic acid hydrochloride, rs-pyrrolidine-3-carboxylic acid hydrochloride
44890816
pyrrolidine-3-carboxylic acid;hydrochloride
C1CNCC1C(=O)O.Cl

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY
Melting Point 30°C to 38°C
Formula Weight 151.59
Percent Purity 95%
Quantity 250 mg
Flash Point >110°C (230°F)
Chemical Name or Material DL-beta-Proline hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'