Learn More
DL-Alanine-3,3,3-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | DL-Alanine-3,3,3-D3 |
| CAS | 53795-94-1 |
| Molecular Formula | C3 D3 H4 N O2 |
| Quantity | 0.5 g |
| Synonym | Alanine-3,3,3-d3 (9 CI, ACI), DL-Alanine-3,3,3-d3 (ZCI), 2-Amino-3,3,3-trideuteriopropanoic acid, DL-(β-2H3)Alanine, DL-[3,3,3-d3]Alanine, β,β,β-Trideutero-DL-alanine, 2-Aminopropanoic-3,3,3-d3 acid, DL-Alanine-3,3,3-D3, DL-Alanine-d3 |
| InChI Formula | InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(N)C(=O)O |
| IUPAC Name | 2-amino-3,3,3-trideuteriopropanoic acid |
| Molecular Weight (g/mol) | 92.1117 |
| Formula Weight | 92.0665 g/mol |
| Show More |