missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DKK2 Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
1834.00 NOK - 4630.00 NOK
Specifications
| Antigen | DKK2 |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17604316
|
Invitrogen™
MA543856 |
20 μL |
1834.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17694306
![]() |
Invitrogen™
MA543855 |
100 μL |
4630.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. Activity of this protein is also modulated by binding to the Wnt co-receptor LDL-receptor related protein 6.Tekniske data
| DKK2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| dickkopf 2; dickkopf 2 homolog; Dickkopf gene 2; dickkopf homolog 1; dickkopf homolog 2; dickkopf homolog 2 (Xenopus laevis); dickkopf homolog 4; dickkopf related protein-2; dickkopf WNT signaling pathway inhibitor 2; dickkopf-2; Dickkopf-related protein 2; DKK2; DKK-2; Dkk4; hDkk-2; mDkk-2; UNQ682/PRO1316 | |
| DKK2 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UBU2 | |
| 27123 | |
| Ile38-Lys258 (Accession Q9UBU2), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| DKK2 |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel