missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-1 Antibody (BHQ-880) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28459JF635
This item is not returnable.
View return policy
Description
Dkk-1 Monoclonal antibody specifically detects Dkk-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Dkk-1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| DKK1 | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 22943 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| BHQ-880 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction