missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKC1 Rabbit anti-Human, Mouse, Rat, Clone: S02-2D8, Novus Biologicals™
Rabbit Monoclonal Antibody
2645.00 NOK - 5365.00 NOK
Specifications
| Antigen | DKC1 |
|---|---|
| Clone | S02-2D8 |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18329206
|
Novus Biologicals
NBP3-14925-25UL |
25 μg |
2645.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18349271
|
Bio-Techne
NBP3-14925-100UL |
100 μg |
5365.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DKC1 Monoclonal antibody specifically detects DKC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| DKC1 | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CBF5, CBF5 homolog, cbf5p homolog, DKC, dyskeratosis congenita 1, dyskerin, Dyskerin, EC 5.4.99, EC 5.4.99.-, FLJ97620, H/ACA ribonucleoprotein complex subunit 4, NAP57, NOLA4dyskerin, Nopp140-associated protein of 57 kDa, Nucleolar protein family A member 4, Nucleolar protein NAP57, snoRNP protein DKC1, XAP101 | |
| A synthetic peptide of human DKC1 (Uniprot#: O60832) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S02-2D8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 1736 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title