Learn More
Wesbart™ Divided Storage Tray
Description
- Use the division guide on the inside cover to check the size of the test tube you wish to store
- As the cells have no spacing in between remember to accommodate for the cap or lid size too
Specifications
Specifications
| Color | Natural |
| Holds | Samples (110) |
| Material | Aluminum |
| Dimensions (L x W x H) | 133 x 555 x 34 mm |
| Product Type | Divided Tray |
| Height (Metric) | 34 mm |
| Array | 5 x 22 |