Description
- Inner door divided and compartmentation
- Internal door divided into 4, 6 or 8 (depending on model) parts and compartmentation of the interior
Specifications
Specifications
| Description | Inner door, divided - Cell therapy compartmentalization, consisting of 4 inner doors and two shelf levels with one partition wall each |
| For Use With (Equipment) | BINDER Series CB and CBF |
| Material | Silicone |
| Type | Access Port |
| Door Style | 4-part Internal Door |
| Features | Internal door divided into 4 parts |