Learn More
Lenz Laborglasintrumente Distribution Tubes With Frit
Description
- Distribution tubes with frit (porosity: 0)
- For specific light solvents
- Made of DURAN™ tubing
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Duran Glass |
| For Use With (Application) | Specific light solvents |