Learn More
Lenz Laborglasinstrumente™ Distilling Links with Liebig Condenser and Head to Claisen
Specifications
Specifications
| For Use With (Application) | For distillation thermometers of 55 mm built-In length |
| Type | Distilling Link |
| Material | Duran Glass |
| Standard Taper Joint | NS 29/32, NS 29/32 |
| Length (Metric) Jacket | 250 mm |