Learn More
Lenz Laborglasintrumente Distilling Links with Liebig Condenser and GL Thread
Product Code. p-7222543
Click to view available options
Standard Taper Joint:
14/23 NS
29/32 NS
Unit Size:
Each
Product Code. 11562324
Brand: Lenz Laborglasintrumente 05050414
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Distilling links with Liebig condenser
  • Outlet with NS cone, water connection with thread GL14
  • With socket NS 14/23 for distillation thermometers of 55 mm built-in length
  • Made of DURAN™ tubing
Specifications
Length (Metric) 400 mm
Material Duran Glass
Quantity 1 Ea.
Standard Taper Joint 29/32 NS