Learn More
Brady™ Dissolvable Paper Labels
Product Code. 17851505
Click to view available options
Length (English):
100 ft.
300 ft.
Width (English):
1.125 in.
2.25 in.
3 in.
4 in.
Unit Size:
1 roll
Product Code. 17851505
Brand: Brady™ 149275
Only null left
Add to basket
Add to basket
Brady™ Dissolvable Paper Labels completely dissolve in warm water within 30 seconds, leaving no adhesive residue behind.
  • Ideal for use in laboratory settings, asset tracking, or general use
  • Easy to remove, saving time during re-labeling process
  • Environmentally responsible
  • Pressure-sensitive adhesive allows for quick and easy application
Compliance
RoHS directive 2011/65/EU
Specifications
Adhesive Type Removable
Coating None
Color White
Finish Matte
Format Continuous
Material Paper
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset tracking, General identification, Laboratory labelling, Warehouse marking
For Use With (Equipment) BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -70°C
Model THT-2250-403-WT
No. per Pack 1 Tape / Roll
Quantity 1
Size 57.15 mm (W) x 91.44 m (L)
Temperature Range (English) -94°F to +212°F
Temperature Range (Metric) -70°C to +100°C
Thickness (English) 0.0030 in.
Thickness (Metric) 0.0762 mm
Dimensions (LxW) 57.15 mm (W) x 91.44 m (L)
Length (English) 100 ft.
Length (Metric) 91.44 m
Width (English) 2.25 in.
Width (Metric) 57.15 mm
Width (English) Printable Area 57.15 mm
Alternate Part Number THT-2250-403-WT