Learn More
Corning™ Dissolvable Microcarriers
Corning™ Dissolvable Microcarriers are designed to simplify cell harvest, separation and concentration processes and are ideal for bioprocess scale-up.
1510.50 NOK - 90402.10 NOK
Specifications
Product Type | Dissolvable Microcarrier |
---|---|
For Use With (Application) | Large-volume adherent cell culture scale-up and cell banking |
Surface Treatment | Corning™ Synthemax II |
Culture Area | 5000 cm2/g |
Material | Polygalacturonic acid (PGA) polymer chains cross linked via calcium ions |
Product Code | Brand | Surface Treatment | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Quantity | Price | Quantity & Availability | ||||
15823895
|
Corning™
4989 |
Corning™ Synthemax II | 10 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15813895
|
Corning™
4988 |
Corning™ Synthemax II | 5 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15803895
|
Corning™
4987 |
Corning™ Synthemax II | 1 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15833895
|
Corning™
4990 |
Corning™ Synthemax II | 100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Offer cell separations that are faster, gentler, and more convenient than other standard commercially available microcarriers
- Comprised of polygalacturonic acid (PGA) polymer chains cross linked via calcium ions pre-coated with either denatured collagen or Corning Synthemax™ II to enhance cell attachment for optimal yield and viability
- Dissolvable within 10 to 20 minutes using EDTA and pectinase for easy separation of cells from the actual microcarriers
- Narrow size distribution (1.02-1.03 g/cm3 density, 200-300μm fully hydrated) and large surface area (5000cm2/g) result in a more homogenous culture
- Optical clarity for visualizing cell morphology
- Presterilized to Sterility Assurance Level (SAL) of 10-6
- Nonpyrogenic
- Applications areas include cell therapy, vaccine production or large-volume adherent cell culture scale-up and cell banking
Specifications
Dissolvable Microcarrier | |
Corning™ Synthemax II | |
Polygalacturonic acid (PGA) polymer chains cross linked via calcium ions |
Large-volume adherent cell culture scale-up and cell banking | |
5000 cm2/g | |
200 to 300 μm fully hydrated |