Learn More
Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves
Shop All Fisherbrand Products
Click to view available options
Size:
X-Small
Small
Medium
Large
Unit Size:
Pack of 50
Description
- 100% nitrile-resistant to a wide range of chemicals and three times more puncture resistant than latex
- Ergonomically designed with textured fingertips for improved grip
- Latex, protein and powder free to eliminate Type I allergies
- 0.10mm thickness
- Non-detectable residual accelerators
- Exceeds the requirements of EN 374 PPE Category III
- Conforms to EN 455 parts 1 and 2
Specifications
Specifications
| Certifications/Compliance | Conforms to EN 455 parts 1 and 2 |
| Color | Purple |
| Disposable | Single-use |
| Latex Free | Yes |
| Material | Nitrile |
| Powdered | Powder Free |
| Product Type | Nitrile Examination Glove |
| Size | X-Small |
| Sterility | Non-sterile |
| Thickness (Metric) | 0.10 mm |
| Show More |