Learn More
Kartell™ Disposable Polystyrene Cell Counter Cup
Product Code. 11373804
Change view
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11373804 25 mL Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11373804 Supplier Kartell™ Supplier No. 0093600
  • ONLINE EXCLUSIVE PRICE
    815.00 NOK
    312.97 NOK / Pack of 200
    Save 502.03 NOK
    61% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Manufactured with clear polystyrene for optimum visibility. Kartell™ Coulter™ Cell Counter Cups are used for specimen analysis with cell counters.
  • Disposable
  • Includes polyethylene caps
Specifications
Quantity 200 Pk.
Capacity (Metric) 25 mL
Color Clear
Product Type Cell Counter Cup
Material Polystyrene
Disposable Single-use
For Use With (Application) For Analysis
For Use With (Equipment) Fr Toa™ and Royco-Hycel™ cell counters
Height (Metric) 56 mm