Learn More
Kartell™ Technicon™ Beckman™, Konelab™ Polystyrene Sample Cups
Description
Manufactured with crystal clear polystyrene for optimum visibility. Kartell™ Technicon™ Beckman™ Konelab™ Sample Cups are used for specimen analysis.
- A range of crystal clear polystyrene auto analyser cups with conical bases
- For Technicon™, Beckman™, Konelab™
- Supplied without caps
- Graduated
Specifications
Specifications
| Quantity | 1000 Pk. |
| Capacity (Metric) | 1.5 mL |
| Color | Crystal Clear |
| Product Type | Sample Cup |
| Material | Polystyrene |
| Disposable | Single-use |
| For Use With (Equipment) | Technicon, Beckman and Konelab analyzers |
| Height (Metric) | 22.6 |