Learn More
3M™ Disposable Maintenance-Free Half Mask Respirator, 4000 Series
Product Code. 12372399
Change view
Click to view available options
Cartridge Type:
FFABEK1P3 R D
Unit Size:
Each
Product Code. Cartridge Type unitSize
12372399 - -
12392399 - -
12302409 - -
12312409 FFABEK1P3 R D Each
11733343 - Each
5 options
Product Code. 12372399
Brand: 3M™ 7000034735
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offers reliable respiratory protection against a range of gases, vapors, and particulates. The 3M Disposable Maintenance-Free Half Mask Respirator can be used until damaged, particulate filters clog, gas filters become saturated or after one month.
Simple, lightweight, well-balanced respirator offers reliable respiratory protection against a range of gases, vapors, and particulates.
  • No parts assembly or maintenance required
  • Unique bonded carbon technology allows the respirator to sit close to the face
  • Low profile design allows the respirator to fit under many other types of protective equipment, providing an undisturbed field of vision and a more comfortable worklife
  • Exhalation valve centrally positioned for effective removal of heat and moisture
  • Can be used until damaged, particulate filters clog, gas filters become saturated or after one month
Respiratory protection against:
  • Fine dusts
  • Oil and water based mists
  • Organic gases or vapours with good warning properties and boiling point above 65°C
Approved to:
  • EN 405:2002
  • Maximum usage limitations:
  • Gases/vapours: APF - 10 x WEL
  • Particulates: APF - 20 x WEL
  • CE marked 