Learn More
Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeve
Product Code. 15346459
Change view
Click to view available options
For Use With (Equipment):
For ground joints NS 14/23
For ground joints NS 24/29
For ground joints NS 45/40
Unit Size:
Pack of 10
Product Code. For Use With (Equipment) unitSize
15346459 For ground joints NS 24/29 Pack of 10
10862231 For ground joints NS 14/23 Pack of 10
10617153 For ground joints NS 45/40 Pack of 10
3 options
Product Code. 15346459
Brand: Lenz Laborglasinstrumente™ 01101524
Only null left
Add to basket
Add to basket
Fits like a second skin without adding bulk
  • Thin walled
  • Outstanding chemical resistance at operating temperatures from -200 to +260 °C
  • Without grip collar, simplifying use of joint clips
Specifications
For Use With (Equipment) For ground joints NS 24/29
Material PTFE
Quantity 10
Temperature Range (Metric) -200 to +260 °C
Thickness 50 μm
Standard Taper Joint No. 24/29
Type Disposable