Learn More
Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeve
Click to view available options
For Use With (Equipment):
For ground joints NS 14/23
For ground joints NS 24/29
For ground joints NS 45/40
Unit Size:
Pack of 10
Description
- Thin walled
- Outstanding chemical resistance at operating temperatures from -200 to +260 °C
- Without grip collar, simplifying use of joint clips
Specifications
Specifications
| For Use With (Equipment) | For ground joints NS 45/40 |
| Material | PTFE |
| Quantity | 10 |
| Temperature Range (Metric) | -200 to +260 °C |
| Thickness | 50 μm |
| Standard Taper Joint No. | 45/40 |
| Type | Disposable |