Learn More
Buerkle™ Disposable Funnel for Powders, LaboPlast™/SteriPlast™
BIOPHARMAGREENER_CHOICE
Product Code. 17590846 Shop All Bürkle Products
Click to view available options
Color:
Blue
White
Material:
Bio-PE
Polystyrene
Polystyrene, Detectable
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Product Code. 17590846
Brand: Buerkle™ 53791041
Only null left
Add to basket
Add to basket
The Disposable Funnel for Powders, LaboPlast™/SteriPlast™ has a short, wide discharge and is suitable for transferring powdered samples and materials, granulates, and suspensions, as well as other liquids.
  • For easy and reliable transferring
  • Wide opening prevents clogging
  • Suitable for use in laboratories and clean rooms or for sampling
  • Clean room manufactured (clean room class 7)
  • Individually packaged and optionally sterilized
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 100 mm
Diameter (Metric) Stem 25 mm
Material Bio-PE
Color White
For Use With (Application) Transferring Powders, Granulates and Suspensions
Sterility Sterile
Product Type Filling Funnel (Powders)