Learn More
Buerkle™ Disposable Funnel for Liquids, LaboPlast™/SteriPlast™
BIOPHARMAGREENER_CHOICE
Product Code. 17580846 Shop All Bürkle Products
Click to view available options
Color:
Blue
White
Material:
Bio-PE
Polystyrene
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Product Code. 17580846
Brand: Buerkle™ 53791040
Only null left
Add to basket
Add to basket
The Disposable Funnel for Liquids, LaboPlast™/SteriPlast™ for reliable filling of liquids, is ideal for filling bottles and other containers with a narrow filler opening.
  • For easy and reliable filling or filtering
  • Suitable for use in laboratories and clean rooms or for sampling
  • Cleanroom manufactured (Class 7)
  • Individually packaged and optionally sterilized
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 100 mm
Diameter (Metric) Stem 25 mm
Material Bio-PE
Height (Metric) 149 mm
Color White
For Use With (Application) Transferring Powders, Granulates and Suspensions
Sterility Sterile
Product Type Filling Funnel (Liquid)