Description
Disposable Blade Carriers
- Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
- Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
- Quick clamp system for high- and low-profile blades
- Adjustable clearance angle
- Moveable knife guard covers the blade entirely in any position
- Knife carrier base included
Specifications
Specifications
| Description | E Type blade carrier for dovetail style units |
| Product Type | Disposable Blade Carrier E |
| For Use With (Equipment) | Dovetail Style Unit |