Learn More
Epredia™ Disposable Base Molds
Product Code. 12510117 Shop All Epredia Products
Change view
Click to view available options
Dimensions (L x W):
30 x 24 mm
37 x 24 mm
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12510117 30 x 24 mm Pack of 500
12520117 37 x 24 mm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12510117 Supplier Epredia™ Supplier No. 58952
  • ONLINE EXCLUSIVE PRICE
    1670.00 NOK
    1483.80 NOK / Pack of 500
    Save 186.20 NOK
    11% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Use Epredia™ Disposable Base Molds with all standard-size cassettes.
  • Cleaning associated with metal molds is not required
  • Transparency facilitates tissue orientation
Ordering Information:
  • Sold in cases of four boxes.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Base Mold
Material Plastic
Dimensions (L x W) 30 x 24 mm