Learn More
CellPath DispoCut™ Board
Product Code. 12826653
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12826653 White Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12826653 Supplier CellPath Supplier No. CGB050253A
  • ONLINE EXCLUSIVE PRICE
    1358.23 NOK
    1074.78 NOK / Pack of 48
    Save 283.45 NOK
    20% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Disposable cutting board designed to increase safety and hygiene in high-risk areas as well as routine cutting areas.
  • Manufactured from 3mm, three ply foam filled board for strength and rigidity
  • Offers high degree of moisture resistance and helps protect work surfaces while ensuring accurate dissection
  • Features metric and imperial scale to assist in macroscopic and photographic recording of specimens
  • Easily disposed of by incineration
Specifications
Color White
Size 148 x 210 mm