Learn More
Honeywell™ Dispensing System and Dip Tube
Product Code. 16871210 Shop All Honeywell Fluka Products
Change view
Click to view available options
For Use With (Equipment):
18 L Drum without Dip Tube
Capacity (Metric):
18 L
Unit Size:
1 pack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Capacity (Metric) Unit Size
16871210 18 L Drum without Dip Tube 18 L 1 pack
16881210 - - -
16891210 - - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16871210 Supplier Honeywell™ Supplier No. 89857H1EA
  • ONLINE EXCLUSIVE PRICE
    3555.00 NOK
    3447.51 NOK / Pack of 1
    Save 107.49 NOK
    3% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Dispensing system and dip tube
TRUSTED_SUSTAINABILITY
Specifications
Product Type Returnable Container Accessory
Capacity (Metric) 18 L
For Use With (Equipment) 18 L Drum without Dip Tube
Quantity 1
Weight (Metric) 0.675 Kg