Learn More
BRAND™ Tilt Measure Dispenser with Bottle
Product Code. 10399851 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
10399851 50 mL Each
10138371 10 mL Each
10257971 100 mL Each
10148371 20 mL Each
10128371 5 mL Each
10064151 25 mL Each
6 options
Product Code. 10399851
Brand: BRAND™ 43028
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Particularly well-suited for dispensing suspensions. Comes complete with 1000mL soda-lime glass reservoir bottle.
  • Dispenser head made from Borosilicate glass (Boro 3.3)
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • Particularly well-suited for dispensing suspensions
  • Complete with 1000mL soda-lime glass reservoir bottle with 29/32 neck size
TRUSTED_SUSTAINABILITY
Specifications
Product Type Top Tilt Pipette Dispenser
Color Clear
Material Borosilicate Glass
Volume (Metric) 50 mL
Includes 1000mL soda-lime glass reserve bottle