Learn More
Masterflex™ Straight Tubing Connector
Product Code. 15531295
Change view
Click to view available options
Diameter (English) Inner:
0.19 in.
0.25 in.
0.31 in.
0.38 in.
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
15531295 0.19 in. Pack of 10
12683606 0.25 in. Pack of 10
15511295 0.31 in. Pack of 10
15521295 0.38 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15531295 Supplier Masterflex™ Supplier No. 4062257
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
With barbed fittings.
  • Made from polypropylene
  • Ethylene Oxide/Gamma sterilized
Specifications
Diameter (English) Inner 0.19 in.
Diameter (Metric) Inner 0.48 cm
Material Polypropylene
Sterilization Method ETO or Gamma
Includes Barbed fittings
Product Type Straight Tubing Connector