Learn More
Masterflex™ Straight Tubing Connector
Product Code. 15521295 Shop All Masterflex Products
Change view
Click to view available options
Diameter (English) Inner:
0.19 in.
0.25 in.
0.31 in.
0.38 in.
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Unit Size
15521295 0.38 in. Pack of 10
15531295 0.19 in. Pack of 10
12683606 0.25 in. Pack of 10
15511295 0.31 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15521295 Supplier Masterflex™ Supplier No. 4061002
  • ONLINE EXCLUSIVE PRICE
    303.00 NOK
    254.47 NOK / Pack of 10
    Save 48.53 NOK
    16% Off
Estimated Shipment: 12-02-2027
With barbed fittings.
  • Made from polypropylene
  • Ethylene Oxide/Gamma sterilized
Specifications
Diameter (English) Inner 0.38 in.
Diameter (Metric) Inner 0.97 cm
Material Polypropylene
Sterilization Method ETO or Gamma
Includes Barbed fittings
Product Type Straight Tubing Connector