Learn More
Masterflex™ Straight Tubing Connector
Click to view available options
Diameter (English) Inner:
0.19 in.
0.25 in.
0.31 in.
0.38 in.
Unit Size:
Pack of 10
Description
- Made from polypropylene
- Ethylene Oxide/Gamma sterilized
Specifications
Specifications
| Diameter (English) Inner | 0.25 in. |
| Material | Polypropylene |
| Sterilization Method | ETO or Gamma |
| Includes | Barbed fittings |
| Product Type | Straight Tubing Connector |