Learn More
BRAND™ Dispenser Box for Schulke Wipes
Product Code. 15431945 Shop All Brand Products
Change view
Click to view available options
Dimensions (L x W):
230 x 280 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15431945 230 x 280 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15431945 Supplier BRAND™ Supplier No. 44875
  • ONLINE EXCLUSIVE PRICE
    324.00 NOK
    262.80 NOK / Pack of 1
    Save 61.20 NOK
    18% Off
View all promotions
Estimated Shipment: 25-11-2026
Dispenser Box designed to hold disinfecting fleece wipes.
  • For high-volume use, wipes can be soaked with liquid disinfectant directly in the handy dispenser box
  • Lasts up to 28 days
  • The usage of the surface disinfecting detergent Pursept™ A Xpress are recommended
  • Can hold up to 70 wipes
  • Refillable and firmly closable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dispenser Box
Dimensions (L x W) 230 x 280 mm
For Use With (Equipment) Holds 70 fleece wipes
Includes Empty