Learn More
Disodium NADP (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | Disodium NADP |
| CAS | 24292-60-2 |
| Molecular Formula | C21 H26 N7 O17 P3 . 2 Na |
| Quantity | 1 g |
| Synonym | NAD Sodium Phosphate, Sodium (2R,3R,4R,5R)-2-(6-Amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl Phosphate |
| InChI Formula | InChI=1S/C21H28N7O17P3.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37, |
| SMILES | [Na+].[Na+].NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP(=O)([O-])OP(=O)([O-])OC[C@H]3O[C@H]([C@H](OP(=O)(O)[O-])[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O |
| IUPAC Name | disodium;[(2R,3R,4R,5R)-2-(6-aminopurin-9-yl)-5-[[[[(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]-4-hydroxyoxolan-3-yl] hydrogen phosphate |
| Molecular Weight (g/mol) | 787.369 |
| Formula Weight | 787.039 |