missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium hydrogen phosphate, anhydrous, Scharlab
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Specifications
Specifications
| CAS | 7558-79-4 |
| Quantity | 1 kg |
| Molecular Formula | HNa2O4P |
| MDL Number | MFCD00003496 |
| InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
| SMILES | [Na+].[Na+].OP([O-])([O-])=O |
| IUPAC Name | disodium hydrogen phosphate |
| Molecular Weight (g/mol) | 141.96 |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction