Learn More
Helios Italquartz Dishes for Mahler Bomb Calorimeter, Transparent
Click to view available options
Diameter (Metric):
17 mm
20 mm
25 mm
Height (Metric):
16 mm
20 mm
Unit Size:
Each
Description
- Transparent
- Suitable for use in Mahler bomb calorimeter
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Specifications
| Description | Transparent, 37 mm flare |
| Material | Quartz |
| Shape | Round |
| Type | Evaporating Dish |
| For Use With (Application) | Mahler Bomb Calorimeter |
| Height (Metric) | 20 mm |
| Diameter (Metric) | 17 mm |