Description
- Discharge valves are manufactured of PFA/Boro 3.3/ceramic components. The Dispensette™ S discharge valve has a platinum-iridium valve spring; the Dispensette™ S Organic discharge valve has a tantalum valve spring.
- Discharge valves for the Dispensette™ S Trace Analysis are available with either a platinum-iridium or tantalum valve spring for specific applications (e.g., Pt-Ir valve for dispensing HF). Available in two or three sizes: one for 1mL and 2mL models; one for 5mL and 10mL models; one for 25mL, 50mL and 100mL models
Specifications
Specifications
| Product Type | Discharge Valve |
| Autoclavable | Autoclavable |
| Color | Translucent |
| Material | ETFE, Borosilicate Glass, Ceramic |
| For Use With (Application) | Dispensing liquid |
| For Use With (Equipment) | Dispensette S Organic, 5 and 10 mL |
| Includes | Discharge Valve Only |