Learn More
Cole-Parmer™ Stuart™ Discharge Tube for Pressmatic Bottle Top Dispenser
Choose a discharge tube for Model PD10P or PD60P Pressmatic Bottle Top Dispenser
Brand: Cole-Parmer™ 99955-60
Additional Details : Weight : 0.01000kg
Description
- For use with a Stuart™ Model PD10P or PD60P Pressmatic Bottle Top Dispensers
- PD10P works with smaller volumes from 2 to 10mL
- PD60P works with volumes between 10 and 60mL
- Compatible with dilute acids, buffers, polar solvents, biological solutions
Specifications
Discharge Tube | |
Stuart PD10P Pressmatic Bottle Top Dispenser |