Learn More
Lovibond™ Comparator Disc
Product Code. 13121681
Change view
Click to view available options
Analytes:
Ammonia
Chlorine
Chlorine (DPD)
Dissolved Oxygen
Hydrazine
Iron
Nitrate
Ortho-phosphate
ph Value
Description:
Disc 2/1B
Disc 2/1E
Disc 2/1H
Disc 2/1J
Disc 3/113
Disc 3/116
Disc 3/142
Disc 3/2 Hypo
Disc 3/40A
Disc 3/40B
Disc 3/40K
Disc 3/70
Disc NAB
Disc NOE
Disc: NOH
Unit Size:
Each
Pack of 2000
Product Code. Analytes Description unitSize
13121681 Nitrate Disc 3/142 Each
13131681 ph Value Disc 2/1J Each
13176030 ph Value Disc 2/1E Each
13186030 ph Value Disc 2/1H Each
13111681 Iron Disc 3/116 Each
13191671 Chlorine (DPD) Disc 3/40A Pack of 2000
13126040 Chlorine Disc 3/40K Each
13101681 Chlorine Disc 3/40B Each
13166030 ph Value Disc 2/1B Each
13116040 Ammonia Disc 3/113 Each
13146040 Chlorine Disc 3/2 Hypo Each
13106040 Ortho-phosphate Disc 3/70 Each
12744527 Ammonia Disc NAB Each
12784527 Dissolved Oxygen Disc NOE Each
12794527 Hydrazine Disc: NOH Each
15 options
Product Code. 13121681
Brand: Lovibond™ 230360
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
  • Easy to use, visual assessment of the sample; simply rotate disc to match color
  • Glass, fade-free standards for longevity of use
  • Shock resistant, scratch resistant
  • Optional lighting units for guaranteed lighting conditions 24 hours/day
  • Optional Nessleriser systems for longer path lengths
  • More than 400 color discs are available for the Comparator 2000+
  • Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Specifications
Analytes Nitrate
Description Disc 3/142
For Use With (Equipment) Comparator 2000+
Range 10-100ppm N
Type Color Comparator Disc