Learn More
VELP Scientifica Disc 1 for Capsules for Autosampler
Product Code. 30003202 Shop All Velp Products
Change view
Click to view available options
Type:
Autosampler Disc
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
30003202 Autosampler Disc Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30003202 Supplier VELP Scientifica Supplier No. A00000420
  • ONLINE EXCLUSIVE PRICE
    8116.20 NOK
    7352.68 NOK / Pack of 1
    Save 763.52 NOK
    9% Off
 Restricted Item
Estimated Shipment: 04-11-2026
30-position disc for capsules for liquids or semi-solid samples (expandable with an additional disc A00000420 - up to 59 samples).
  • Additional 30-position disc for capsules, for liquids or semi-solid samples
  • Suitable for the CN 802 Carbon/Nitrogen Analyser
  • The VELP C/N boasts exceptional efficiency, enabling the highest sample throughput. This is made possible by theelectronic autosampler, capable of processing up to 117 samples at a time.
Specifications
Type Autosampler Disc