Learn More
Brady™ Directional Arrow Tape
Product Code. 12777936
Change view
Click to view available options
Dimensions (LxW):
33 m x 100 mm
33 m x 25 mm
33 m x 50 mm
Color:
Black on Gray
Black on Ochre
Black on Orange
Black on White
White on Black
White on Blue
White on Green
White on Purple
White on Red
Unit Size:
1 roll
Product Code. Dimensions (LxW) Color unitSize
12777936 33 m x 25 mm White on Red 1 roll
12747926 33 m x 100 mm Black on Gray 1 roll
12707936 33 m x 100 mm Black on Orange 1 roll
12797936 33 m x 100 mm Black on White 1 roll
12757916 33 m x 100 mm White on Black 1 roll
12787916 33 m x 100 mm White on Blue 1 roll
12767926 33 m x 100 mm White on Green 1 roll
12737936 33 m x 100 mm White on Purple 1 roll
12767936 33 m x 100 mm White on Red 1 roll
12797926 33 m x 25 mm Black on Gray 1 roll
12707946 33 m x 25 mm Black on White 1 roll
12797916 33 m x 25 mm White on Blue 1 roll
12759664 33 m x 25 mm Black on Ochre 1 roll
12777926 33 m x 25 mm White on Green 1 roll
12717936 33 m x 25 mm Black on Orange 1 roll
12767916 33 m x 25 mm White on Black 1 roll
12747936 33 m x 25 mm White on Purple 1 roll
12757926 33 m x 50 mm Black on Gray 1 roll
12727936 33 m x 50 mm Black on Orange 1 roll
12717946 33 m x 50 mm Black on White 1 roll
12769664 33 m x 50 mm Black on Ochre 1 roll
12777916 33 m x 50 mm White on Black 1 roll
12707926 33 m x 50 mm White on Blue 1 roll
12787926 33 m x 50 mm White on Green 1 roll
12757936 33 m x 50 mm White on Purple 1 roll
12787936 33 m x 50 mm White on Red 1 roll
26 options
Product Code. 12777936
Brand: Brady™ 275102
Only null left
Add to basket
Add to basket
Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
  • Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
Specifications
Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Directional Arrow Tape
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Red
Finish Gloss
Format Tape
Material Polyester
Product Type Pipe Marker
For Use With (Application) Pipe and valve marking
No. per Pack 1300 Markers/Roll
Quantity 1300 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (LxW) 33 m x 25 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 0.98 in.
Width (Metric) 25 mm
Alternate Part Number ARROWTAPE:WHITE/RED25MMX33M