missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Direct Red 75, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| CAS | 2829-43-8 |
| Molecular Formula | C33H22N8Na4O15S4 |
| Synonym | Benzo Fast Pink 2 BL, Sirius Rose BB |
| InChI Key | ZXRQHEAKVNYRIS-UHFFFAOYSA-J |
| SMILES | C1=CC(=C(C2=C(C=C(C=C21)S(=O)(=O)O)[O-])N=NC3=C(C=C(C=C3)N=C(NC4=CC(=C(C=C4)N=NC5=C(C=CC6=CC(=CC(=C65)[O-])S(=O)(=O)[O-])N)S(=O)(=O)O)[O-])S(=O)(=O)O)N.[Na+].[Na+].[Na+].[Na+] |
| IUPAC Name | tetrasodium;6-amino-5-[[4-[[N-[4-[(2-amino-8-oxido-6-sulfonaphthalen-1-yl)diazenyl]-3-sulfophenyl]-C-oxidocarbonimidoyl]amino]-2-sulfophenyl]diazenyl]-4-oxidonaphthalene-2-sulfonate |
| Molecular Weight (g/mol) | 990.779 |
| PubChem CID | 101590378 |
| Formula Weight | 902.853 |
| Quantity | 10 g |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction