Learn More
Dipropylene Glycol, Sigma-Aldrich™
Specifications
Specifications
| CAS | 25265-71-8 |
| Melting Point | -32°C |
| Density | 1.02 g/cm3 at 20°C |
| Boiling Point | 231°C to 235°C/1013 hPa |
| MDL Number | MFCD29035085 |
| Quantity | 1 L |
| Synonym | Dipropylene glycol, Dipropylene glycol |
| Molecular Weight (g/mol) | 134.17 |
| Grade | Research |